Learn More
Medchemexpress LLC Gol-NTR | 2968461-58-5 | C24H16F3N3O4 | 100 MG
SDP
Catalog No. 5000373148
Encompass_Preferred
Gol-NTR is a Golgi-targetable probe activated by Nitroreductase (NTR), designed for highly selective and sensitive visualization of acute lung injury (ALI) and assessment of its repair function. It has a low detection limit of 54.8 ng/mL, making it suitable for monitoring and assessing the research response of sepsis-induced ALI, as well as for in vitro fluorescent labeling and in vivo imaging studies.
  • High selectivity and sensitivity
  • Low detection limit of 54.8 ng/mL
  • Visualizes acute lung injury (ALI)
  • Assesses ALI repair function
  • Monitors sepsis-induced ALI response
  • Suitable for in vitro fluorescent labeling of NTR in A549 cells
  • Applicable for in vivo imaging in animal models
Catalog No. 50-003-73148 Supplier Medchemexpress LLC Supplier No. HY151537100MG
Gol-NTR is a Golgi-targetable probe activated by Nitroreductase (NTR), designed for highly selective and sensitive visualization of acute lung injury (ALI) and assessment of its repair function. It has a low detection limit of 54.8 ng/mL, making it suitable for monitoring and assessing the research response of sepsis-induced ALI, as well as for in vitro fluorescent labeling and in vivo imaging studies.
  • High selectivity and sensitivity
  • Low detection limit of 54.8 ng/mL
  • Visualizes acute lung injury (ALI)
  • Assesses ALI repair function
  • Monitors sepsis-induced ALI response
  • Suitable for in vitro fluorescent labeling of NTR in A549 cells
  • Applicable for in vivo imaging in animal models