Learn More
Medchemexpress LLC Gol-NTR | 2968461-58-5 | C24H16F3N3O4 | 25 MG
SDP
Catalog No. 5000374494
Encompass_Preferred
Gol-NTR is a Golgi-targetable probe characterized by high selectivity and sensitivity. It is activated by Nitroreductase (NTR) and is capable of visualizing acute lung injury (ALI) and its repair function. The probe has a low detection limit of 54.8 ng/mL. It is suitable for research aimed at monitoring and assessing the response to sepsis-induced ALI.
  • High selectivity and sensitivity
  • Nitroreductase (NTR)-activated
  • Visualizes acute lung injury (ALI) and its repair function
  • Low detection limit of 54.8 ng/mL
  • Suitable for monitoring and assessing response to sepsis-induced ALI
  • Golgi-targetable probe
Catalog No. 50-003-74494 Supplier Medchemexpress LLC Supplier No. 25MGHY151537
Gol-NTR is a Golgi-targetable probe characterized by high selectivity and sensitivity. It is activated by Nitroreductase (NTR) and is capable of visualizing acute lung injury (ALI) and its repair function. The probe has a low detection limit of 54.8 ng/mL. It is suitable for research aimed at monitoring and assessing the response to sepsis-induced ALI.
  • High selectivity and sensitivity
  • Nitroreductase (NTR)-activated
  • Visualizes acute lung injury (ALI) and its repair function
  • Low detection limit of 54.8 ng/mL
  • Suitable for monitoring and assessing response to sepsis-induced ALI
  • Golgi-targetable probe