Learn More
Medchemexpress LLC Gol-NTR | 2968461-58-5 | C24H16F3N3O4 | 5 MG
SDP
Catalog No. 5000372933
Encompass_Preferred
Gol-NTR is a Golgi-targetable probe characterized by its high selectivity and sensitivity. It is activated by Nitroreductase (NTR) and is capable of visualizing acute lung injury (ALI) as well as its repair function. The probe exhibits a low detection limit of 54.8 ng/mL, making it suitable for research focused on monitoring and assessing the response to sepsis-induced ALI.
  • High selectivity and sensitivity
  • Golgi-targetable
  • Nitroreductase (NTR)-activated
  • Capable of visualizing acute lung injury (ALI)
  • Capable of visualizing ALI repair function
  • Low detection limit of 54.8 ng/mL
  • Suitable for monitoring and assessing response to sepsis-induced ALI
Catalog No. 50-003-72933 Supplier Medchemexpress LLC Supplier No. HY1515375MG
Gol-NTR is a Golgi-targetable probe characterized by its high selectivity and sensitivity. It is activated by Nitroreductase (NTR) and is capable of visualizing acute lung injury (ALI) as well as its repair function. The probe exhibits a low detection limit of 54.8 ng/mL, making it suitable for research focused on monitoring and assessing the response to sepsis-induced ALI.
  • High selectivity and sensitivity
  • Golgi-targetable
  • Nitroreductase (NTR)-activated
  • Capable of visualizing acute lung injury (ALI)
  • Capable of visualizing ALI repair function
  • Low detection limit of 54.8 ng/mL
  • Suitable for monitoring and assessing response to sepsis-induced ALI