Learn More
GOLANDER LLC Golander Pump BQ80S Peristaltic Pump, 0.005-64 mL/min, Variable-Speed, 1 ea
Catalog No. NC3934720
Encompass_Preferred
The BQ80S is a portable, microflow variable-speed peristaltic pump designed for precise fluid delivery. It features a versatile flow range of 0.005 to 64 mL/min and supports time-based dispensing modes. Engineered for industrial and laboratory integration, it includes an RS485 MODBUS interface for external control via PC, HMI, or PLC.
  • Flow range from 0.005 to 64 mL/min with 0.1 rpm speed resolution.
  • Speed accuracy of 0.5 percent.
  • Reversible direction, start/stop, and adjustable speed controls.
  • Time dispense function for repetitive dosing based on duration.
  • Optically isolated external control signals for speed, direction, and start/stop.
  • Dust-proof and moisture-proof conformal coated circuit board.
  • Four-digit LED display with membrane keypad operation.
  • Embedded plastic housing design.
Catalog No. NC3934720 Supplier GOLANDER LLC Supplier No. BQ80SDW103
The BQ80S is a portable, microflow variable-speed peristaltic pump designed for precise fluid delivery. It features a versatile flow range of 0.005 to 64 mL/min and supports time-based dispensing modes. Engineered for industrial and laboratory integration, it includes an RS485 MODBUS interface for external control via PC, HMI, or PLC.
  • Flow range from 0.005 to 64 mL/min with 0.1 rpm speed resolution.
  • Speed accuracy of 0.5 percent.
  • Reversible direction, start/stop, and adjustable speed controls.
  • Time dispense function for repetitive dosing based on duration.
  • Optically isolated external control signals for speed, direction, and start/stop.
  • Dust-proof and moisture-proof conformal coated circuit board.
  • Four-digit LED display with membrane keypad operation.
  • Embedded plastic housing design.