Learn More
Strem, An Ascensus Company CAS# 7440-57-5. 10g. Gold 1% on zinc oxide granulate (AUROlite™ Au/ZnO). MFCD00003436
SDP
Catalog No. NC1712678
Encompass
CAS# 7440-57-5. 10g. Gold 1% on zinc oxide granulate (AUROlite™ Au/ZnO). MFCD00003436. Molecular Weight: . Molecular Formula: . Color/form: dark purple granulate 1-2mm dia. Strem# 79-0170. http://www.strem.com/catalog/v/79-0170/
Catalog No. NC1712678 Supplier Strem, An Ascensus Company Supplier No. 79017010G
CAS# 7440-57-5. 10g. Gold 1% on zinc oxide granulate (AUROlite™ Au/ZnO). MFCD00003436. Molecular Weight: . Molecular Formula: . Color/form: dark purple granulate 1-2mm dia. Strem# 79-0170. http://www.strem.com/catalog/v/79-0170/