Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold

Catalog No. 503094231 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-309-4231 5 Ea.
50-309-5738 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-309-4231 Supplier Sigma Aldrich Supplier No. GF452202835EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold

Specifications

CAS 7440-57-5
Melting Point 1063°C
Boiling Point 2808°C
Density 19.3 g/mL (at 25°C)
Quantity 5 Ea.
Molecular Formula Au
Linear Formula Au
MDL Number MFCD00003436
Synonym AU000110
Molecular Weight (g/mol) 196.97
Percent Purity 99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.