Learn More
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Concentration or Composition Notes | 1,000ppm Au |
| CAS Min % | 93.0, 4.13, 0.1 |
| CAS Max % | 97.0, 4.3, 0.1 |
| Equivalence | 1 mL = 1 mg Au |
| Physical Form | Liquid |
| Boiling Point | 100°C |
| Type | Single-element Standard Solution |
| Quantity | 500 mL |
| Grade | High Purity |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
