Learn More
Bellco Glass, Inc. Gold Alum Seal PTFE/RR,11mm PTFE/Red Rubber
SDP
Catalog No. 501945413
Encompass_Preferred
Larger opening provides increased target area for needle penetration. Available in a variety of colors for easy sample identification. MADE IN THE USA. Size: 11mm Septa: clear ptfe or red rubber
Catalog No. 50-194-5413 Supplier Bellco Glass, Inc. Supplier No. 207005200
Larger opening provides increased target area for needle penetration. Available in a variety of colors for easy sample identification. MADE IN THE USA. Size: 11mm Septa: clear ptfe or red rubber