Learn More
Strem, An Ascensus Company CAS# 1121960-90-4. 100mg. Bis(trifluoromethanesulfonyl)imide(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)gold(I), 98%. MFCD21608485
SDP
Catalog No. 502045409
Encompass
CAS# 1121960-90-4. 100mg. Bis(trifluoromethanesulfonyl)imide(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)gold(I), 98%. MFCD21608485. Molecular Weight: 887.64. Molecular Formula: C28H35AuF6NO6PS2. Color/form: white to off-white solid Strem# 79-0230. http://www.strem.com/catalog/v/79-0230/
Catalog No. 50-204-5409 Supplier Strem, An Ascensus Company Supplier No. 790230100MG
CAS# 1121960-90-4. 100mg. Bis(trifluoromethanesulfonyl)imide(2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl)gold(I), 98%. MFCD21608485. Molecular Weight: 887.64. Molecular Formula: C28H35AuF6NO6PS2. Color/form: white to off-white solid Strem# 79-0230. http://www.strem.com/catalog/v/79-0230/