Learn More
Newcomer Supply Gold Chloride 0.2%, Aqueous, 100 ml
SDP
Catalog No. NC2101893
Encompass
Use: Gordon & Sweets Reticulum, Fontana-Masson Argentaffin and GMS Stains. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2101893 Supplier Newcomer Supply Supplier No. 11286A
Use: Gordon & Sweets Reticulum, Fontana-Masson Argentaffin and GMS Stains. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.