Learn More
Newcomer Supply Gold Chloride 1%, Aqueous, 100 ml
SDP
Catalog No. NC0779312
Encompass
Use: Gordon & Sweets and Gomori Basement Membrane, Fontana-Masson Argentaffin and GMS Stains. Can be diluted to 0.25%, 0.2% & 0.1%. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC0779312 Supplier Newcomer Supply Supplier No. 1129A
Use: Gordon & Sweets and Gomori Basement Membrane, Fontana-Masson Argentaffin and GMS Stains. Can be diluted to 0.25%, 0.2% & 0.1%. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.