Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Chloride Solution, 100 mL, G-Biosciences
SCI_ED

Catalog No. S001103
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001103 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001103 Supplier G-Biosciences Supplier No. 7862234
Add to Cart
Add to Cart

Gold Chloride Solution (0.1%) is a laboratory grade reagent essentially used in Reticulum Staining process.

  • Type: Laboratory grade
  • Composition: Gold Chloride Solution (0.1%)
  • State: Liquid
  • Color: Transparent Yellow
  • Odor: Odorless
  • Application: Histology

Specifications

Color Transparent Yellow
Form Liquid
Quantity 100 mL
Product Type Gold Chloride Solution (0.1%)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.