Learn More
Electron Microscopy Sciences Gold Coated Specimen Discs for AFM, 20 mm
SDP
Catalog No. 501930049
Encompass_Preferred
Magnetic stainless steel gold coated specimen discs have smooth edges and consistently flat surfaces to ensure secure holding. Specimens can be mounted directly on the metal discs with double sided tape, epoxy or conductive paste. If a mica specimen support is needed, the mica discs can be easily mounted on the metal support discs.
These discs offer excellent resistance to electromigration, high electrical and thermal conductivity, and high temperatures making them excellent substrates for mounting specimens. Packaged in a polystyrene disc storage box.
Diameter: 20 mm
Catalog No. 50-193-0049 Supplier Electron Microscopy Sciences Supplier No. 7501120
Magnetic stainless steel gold coated specimen discs have smooth edges and consistently flat surfaces to ensure secure holding. Specimens can be mounted directly on the metal discs with double sided tape, epoxy or conductive paste. If a mica specimen support is needed, the mica discs can be easily mounted on the metal support discs.
These discs offer excellent resistance to electromigration, high electrical and thermal conductivity, and high temperatures making them excellent substrates for mounting specimens. Packaged in a polystyrene disc storage box.
Diameter: 20 mm