Learn More
Abcam Gold Conjugation Kit (10 nm, 20 OD), 3 x 2.5UG
Catalog No. NC2241147
Encompass_Preferred
10nm Gold conjugation / labeling in < 4 hrs with 30 secs hands-on time using Gold Conjugation Kit (10 nm, 20 OD) ab201808. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2241147 Supplier Abcam Supplier No. AB2018083X2.5UG
10nm Gold conjugation / labeling in < 4 hrs with 30 secs hands-on time using Gold Conjugation Kit (10 nm, 20 OD) ab201808. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement