Learn More
Abcam Gold Conjugation Kit (40nm, 20 OD), 1 x 10UG
Catalog No. 502582953
Encompass_Preferred
The Abcam Gold Conjugation Kit (ab154873) offers: - Rapid Conjugation: allows for gold conjugation or labeling in less than 4 hours with only 30 seconds of hands-on time. - High Recovery: It ensures 100% antibody recovery, meaning no loss of valuable antibodies. - Stability and Efficiency: The gold nanoparticles have a protective surface coat that provides unrivaled colloidal stability, and the conjugation reaction is initiated simply by reconstituting the freeze-dried nanoparticle
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2953 Supplier Abcam Supplier No. AB1548731X10UG
The Abcam Gold Conjugation Kit (ab154873) offers: - Rapid Conjugation: allows for gold conjugation or labeling in less than 4 hours with only 30 seconds of hands-on time. - High Recovery: It ensures 100% antibody recovery, meaning no loss of valuable antibodies. - Stability and Efficiency: The gold nanoparticles have a protective surface coat that provides unrivaled colloidal stability, and the conjugation reaction is initiated simply by reconstituting the freeze-dried nanoparticle
The product is subject to the following: Abcam Restricted Use Statement