Learn More
Abcam Gold Conjugation Kit (80nm, 20 OD), 3 x 0.5UG
Catalog No. 502582949
Encompass_Preferred
80nm Gold conjugation / labeling in < 4 hrs with 30 secs hands-on time using Gold Conjugation Kit (80nm, 20 OD) ab154876. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2949 Supplier Abcam Supplier No. AB1548763X0.5UG
80nm Gold conjugation / labeling in < 4 hrs with 30 secs hands-on time using Gold Conjugation Kit (80nm, 20 OD) ab154876. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement