Learn More
NANOPROBES INC GoldEnhance EM Plus, 8 mL
Catalog No. NC1115289
Encompass
New, improved gold developer for EM. Comes In four solutions, to be mixed just before use. 8 mL total, enough for 200 grids (25 microliters/grid).
Catalog No. NC1115289 Supplier NANOPROBES INC Supplier No. 21148ML
New, improved gold developer for EM. Comes In four solutions, to be mixed just before use. 8 mL total, enough for 200 grids (25 microliters/grid).