Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Germanium foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7050254
Change view
Click to view available options
Quantity:
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36313BS 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA36313BS Supplier Thermo Scientific Chemicals Supplier No. 036313BS

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Germanium foil
Name Note 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Quantity 10 cm
Odor Odorless
Weight ≈6.45g/10cm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
Percent Purity 99.997%
Grade Premion
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.