Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Germanium slug, 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length, Premion™, 99.998% (metals basis)

Catalog No. AA42983KM
Change view
Click to view available options
Quantity:
25 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42983KM 25 Pc.
AA42983KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42983KM Supplier Thermo Scientific Chemicals Supplier No. 042983KM
Add to Cart

missing translation for 'validMainframeMsg'

Gold Germanium slug is used in scientific research and development activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Germanium slug
Name Note 3.175mm (0.125 in.) dia. x≈3.175mm (0.125 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Density 0.37 g/cm3
Quantity 25 Pc.
Odor Odorless
Weight ≈0.4g/slug
Form Slug
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
Solubility Information Insoluble in water.
Percent Purity 99.998%
Grade Premion
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.