Learn More
Strem, An Ascensus Company CAS# 10294-29-8. 1g. Gold(I) chloride, 97% (99.99%-Au) PURATREM. MFCD00046175
SDP
Catalog No. 5090113409
Encompass
CAS# 10294-29-8. 1g. Gold(I) chloride, 97% (99.99%-Au) PURATREM. MFCD00046175. Molecular Weight: 232.42. Molecular Formula: AuCl. Color/form: yellow pwdr. Strem# 79-1900. http://www.strem.com/catalog/v/79-1900/
Catalog No. 50-901-13409 Supplier Strem, An Ascensus Company Supplier No. 7919001G
CAS# 10294-29-8. 1g. Gold(I) chloride, 97% (99.99%-Au) PURATREM. MFCD00046175. Molecular Weight: 232.42. Molecular Formula: AuCl. Color/form: yellow pwdr. Strem# 79-1900. http://www.strem.com/catalog/v/79-1900/