Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold ICP Standard, 10,000 ppm Au in 5% HCl, Ricca Chemical
SDP

Catalog No. PAU10KH50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PAU10KH-50 50 mL Amber Glass Bottle
PAU10KH-100 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PAU10KH-50 Supplier Ricca Chemical Company Supplier No. PAU10KH50
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-57-5 , 7647-01-0 , 7732-18-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 10,000ppm Au
CAS Min % 83.0, 10.8, 0.83
CAS Max % 87.0, 11.25, 0.87
Equivalence 1 mL = 10 mg Au
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 50 mL
Chemical Name or Material Gold ICP Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.