Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold(III) chloride

Catalog No. 379948250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-994-8250MG 250 mg
37-994-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-994-8250MG Supplier Sigma Aldrich Supplier No. 379948250MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The structure of gold chloride is monoclinic in nature. It sublimes at elevated temperatures.Nanocrystalline superlattices in gold colloidal solutions have been prepared by ligand-induction using AuCl3 reduced with sodium borohydride. Gold chloride may be used to prepare gold cages which may be potentially useful in surface plasmonics , surface- enhanced Raman spectroscopy, drug delivery, contrast-enhanced optical imaging and photothermal therapy.

Specifications

CAS 13453-07-1
Molecular Formula AuCl3
Linear Formula AuCl3
MDL Number MFCD00014172
Quantity 250 mg
Synonym Trichlorogold
Molecular Weight (g/mol) 303.33
Percent Purity ≥99.99% trace metals basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.