Learn More
Medchemexpress LLC Gold(III) chloride hydrate | 27988-77-8 | 99.95% | HAuCl4·XH2O | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW073196500MG
Encompass_Preferred
Gold(III) chloride hydrate is a hydrated tetrachloroauric(III) acid used as a gold precursor in research and materials chemistry, including the preparation of gold nanoparticles and Au-containing catalysts. It is supplied as a hydrated salt with a specified gold basis and is intended for laboratory use.
  • Used as a precursor for gold nanoparticle synthesis.
  • Applicable for preparation of Au-based catalysts and photocatalysts.
  • Provided as a hydrated tetrachloroauric(III) salt (hydrated form).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-10820