Learn More
Medchemexpress LLC Gold(III) chloride hydrate | 27988-77-8 | MFCD00149903 | 50.0% | 339.79 g/mol (anhydrous basis) | HAuCl4·xH2O | 500 MG

Supplier: Medchemexpress LLC HYW073196A500MG
Gold(III) chloride hydrate is a hydrated tetrachloroauric acid supplied as a solid gold precursor for synthesis and analytical use. The material is specified as approximately 50% Au by basis; consult the SDS for handling, storage, and safety information.
- Cas number 27988-77-8.
- Chemical formula HAuCl4·xH2O.
- Approximately 50.0% Au by basis.
- Solid form; store in a cool, dry, ventilated place.
- Sds available for detailed handling and storage instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.