Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold(III) chloride hydrate

Catalog No. 507905G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-790-5G 5 g
50-790-1G 1 g
50-790-250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-790-5G Supplier Sigma Aldrich Supplier No. 507905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold (III) chloride hydrate,also known as chloroauric acid, is used as precursor to synthesize gold nanoparticles. It also exihibits very high catalytic activity.Used as a catalyst in asymmetric aldol reactions, diboration of vinylarenes, dehydrogenative dimerizations of trialkystannanes, carbonylation of olefins and amines, C-C, C-O bond couplings, oxidation of sulfides, selective oxidation of thioethers.

Specifications

CAS 27988-77-8
Density 3.9 g/mL (at 25°C)
Molecular Formula HAuCl4 -+ xH2O
Linear Formula HAuCl4 -+ aq
Quantity 5 g
MDL Number MFCD00149903
Synonym Auric chloride; Chloroauric acid; Hydrogen tetrachloroaurate(III) hydrate; Tetrachloroauric(III) acid
Molecular Weight (g/mol) 339.79
Percent Purity ∼50%Au basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.