Learn More
Medchemexpress LLC Gold III chloride t 100mg
SDP
Catalog No. 5000205763
Encompass_Preferred
Gold (III) chloride trihydrate is a reducing agent Gold (III) chromium trihydrate can be used for chemical synthesis of gold nanoparticles (NP) and spirochromene derivatives[1][2]
Catalog No. 50-002-05763 Supplier Medchemexpress LLC Supplier No. HYW094581100MG
Gold (III) chloride trihydrate is a reducing agent Gold (III) chromium trihydrate can be used for chemical synthesis of gold nanoparticles (NP) and spirochromene derivatives[1][2]