Learn More
Medchemexpress LLC Gold III chloride t 500mg
SDP
Catalog No. 5000206166
Encompass_Preferred
Gold (III) chloride trihydrate is a reducing agent Gold (III) chromium trihydrate can be used for chemical synthesis of gold nanoparticles (NP) and spirochromene derivatives[1][2]
Catalog No. 50-002-06166 Supplier Medchemexpress LLC Supplier No. HYW094581500MG
Gold (III) chloride trihydrate is a reducing agent Gold (III) chromium trihydrate can be used for chemical synthesis of gold nanoparticles (NP) and spirochromene derivatives[1][2]