Alfa Aesar™ Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min

Manufacturer: Alfa Aesar™ 1216106

Quantity 5g

 View more versions of this product

Catalog No. AA1216106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1303-58-8
    Formula Weight 441.93
    MDL Number MFCD00014173
    Merck Index 14,4524
    Chemical Name or Material Gold(III) oxide
    Grade Premion™

    Specifications

    Formula Weight 441.93
    Solubility Information Insoluble in water; Soluble in HCl, HNO3, and in NaCN solution
    Odor Odorless
    Physical Form Powder
    Merck Index 14,4524
    Color Brown
    Melting Point 150°C (dec.)
    TSCA TSCA
    MDL Number MFCD00014173
    CAS 1303-58-8
    Quantity 5g
    Concentration Au 88.6% min.
    Chemical Name or Material Gold(III) oxide
    Assay 99.99%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit