Learn More
TA Instruments Gold Lids, DSC Gold Lids, Use with Gold Sample Pans, -180°C to 725°C, Secure Sealing, High Thermal Stability, 10 Pack
Catalog No. 5000580841 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Gold Lids are DSC gold lids designed for use with gold sample pans (P/N: 900866.901). They are suitable for experiments over -180°C to 725°C, providing secure sealing and high thermal stability for reliable DSC measurements.
Catalog No. 50-005-80841 Supplier TA Instruments Supplier No. 900868.901
Waters TA Instruments Gold Lids are DSC gold lids designed for use with gold sample pans (P/N: 900866.901). They are suitable for experiments over -180°C to 725°C, providing secure sealing and high thermal stability for reliable DSC measurements.