Learn More
TA Instruments Gold Lids Hermetic, DSC Gold Hermetic Lids, High Pressure Materials, up to 6 Atm, -180°C to 725°C, Use with Gold Hermetic Sample Pans, 10 Pack
Catalog No. 5000580647 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Gold Lids, Hermetic are DSC gold hermetic lids designed for evaluating materials under higher self-generated pressures up to 6 atm (600 kPa gage). They have a temperature range of -180°C to 725°C and are used with gold hermetic sample pans (P/N: 900871.901).
Catalog No. 50-005-80647 Supplier TA Instruments Supplier No. 900872.901
Waters TA Instruments Gold Lids, Hermetic are DSC gold hermetic lids designed for evaluating materials under higher self-generated pressures up to 6 atm (600 kPa gage). They have a temperature range of -180°C to 725°C and are used with gold hermetic sample pans (P/N: 900871.901).