Learn More
Sigma Aldrich Fine Chemicals Biosciences Gold microfoil, disks, 10mm, thinness 0.25mum, specific density 483mug/cm2, permanent mylar 3.5mum support, 99.99+% | 7440-57-5 | MFCD00003436 | 1EA
Catalog No. 501851344
Encompass
Gold microfoil, disks, 10mm, thinness 0.25mum, specific density 483mug/cm2, permanent mylar 3.5mum support, 99.99+% | Purity: 99.99+ % | Mol Wt: 196.97 | 7440-57-5 | MFCD00003436 | 1EA
Catalog No. 501851344 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF760097541EA
Gold microfoil, disks, 10mm, thinness 0.25mum, specific density 483mug/cm2, permanent mylar 3.5mum support, 99.99+% | Purity: 99.99+ % | Mol Wt: 196.97 | 7440-57-5 | MFCD00003436 | 1EA