Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold, nano-urchins

Catalog No. 79775825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-775-825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-775-825ML Supplier Sigma Aldrich Supplier No. 79775825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles are extensively used in biomedical research due to their biocompatibility, facile conjugation to biomolecules and unique optical properties arising from surface plasmon resonance. The presence of spiked nanoantenna-type shapes in Nano-Urchins enhances the plasmonic extinction resonance effect, which improves the optical limiting effects.

Specifications

Quantity 25 mL
Molecular Formula Au
Linear Formula Au
Synonym Gold nanoparticles; Non-functionalized gold nanoparticles
Molecular Weight (g/mol) 196.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.