Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 7655381ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-553-81ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-553-81ML Supplier Sigma Aldrich Supplier No. 7655381ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Carboxylic acid functionalized GNPs forms conjugates with various protein molecules due to the electrostatic attraction between negatively charged carboxylate groups and positively charged amino groups of protein.The carboxylic acid group binds with various proteins and are used in targeted drug delivery and gene therapy.

Specifications

Density 1.00 g/cm3
Quantity 1 mL
Synonym Gold nanoparticles COOH functionalized; Au NP COOH; Gold Colloid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.