Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 7652791ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-527-91ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-527-91ML Supplier Sigma Aldrich Supplier No. 7652791ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles (AuNPs) ranging from 2-200nm in diameter find applications in diverse fields. They show excellent biocompatibility and attractive physicochemical properties. Poly (ethylene) glycol (PEG) surface coatings on gold nanoparticles help in the reduction of protein adsorption, reduce nonspecific interactions with cells and greatly improve the pharmokinetics of these nanoparticles and reduce aggregation of the nanoparticles. PEG layer density depends inversely on the AuNP curvature. Denser coating was possible on smaller AuNPs. AuNPs coated with PEG 5000 exhibited highest colloidal stability. Amine functionalized GNPs which have Poly Ethylene Glycol (PEG) as the ligand or carrier molecule forms a conjugate with siRNA and is extremely useful in RNAi technology. Since it adheres to cell membranes, it may find use in cellular and intracellular targeting in targeted drug delivery applications and also used in biodistribution studies.Several other specific applications have been listed below:Since AuNPs adhere to cell membranes, they can be used in cellular and intracellular targeting in targeted drug delivery applications and may also be used in biodistribution studiesAuNPs may also be used in photothermal therapy and radiotherapy.Proposed applications of gold nanoparticles are as:drug delivery agents for cancer therapy, coatings, plastics, nanowire, nanofiber, textiles, catalyst applications. They may also be used in carrying siRNA which acts against human prostate carcinoma cells by inhibiting a specific cancer gene.

Specifications

Density 1.00 g/cm3
Quantity 1 mL
Synonym Gold nanoparticles NH2 functionalized; Au NP NH2; Gold Colloid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.