Learn More
Sigma Aldrich Gold nanoparticles
Catalog No. 9004781EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
90-047-81EA 1 Ea.
1 options
Catalog No. 90-047-81EA Supplier Sigma Aldrich Supplier No. 9004781EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
NHS ester functionalized gold nanoparticles conjugation kit (50 nm) can be used in the surface modification of indium tin oxide (ITO) electrodes for potential usage in electrocatalysis, and electrochemical sensing.AuNPs may be used to prepare SARS-CoV-2 conjugate for COVID-19 detection using lateral flow immunoassay technique. It can be potentially used in immobilization of antibodies to improve the sensitivity of optical fiber sensors for the screening of COVID-19. It can also be deposited on the titanium electrode for the formation of a miniaturized device for the electrochemical detection of DNA hybridization.Product of CytoDiagnostics, Inc.
Specifications
Quantity 1 Ea.
Synonym NHS-activated gold nanoparticles