Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 80894605ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-894-605ML 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-894-605ML Supplier Sigma Aldrich Supplier No. 8089460.5ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles PEG 5000 biotin terminatedGold nanoparticles 80 nm diameter, biotin terminated, PEG 5000 coated, OD 50, dispersion in H2O can be used as a theranostic agent for potential usage in cancer therapy and treatment of COVID-19. It can also be used in the detection of pathogens for biosensing applications. AuNPs may be used to prepare SARS-CoV-2 conjugate for COVID-19 detection using lateral flow immunoassay technique.Product of Cytodiagnositcs, inc

Specifications

Density 1.00 g/cm3
Quantity 0.5 mL
Synonym Biotin Gold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.