Learn More
Sigma Aldrich Gold nanoparticles
Catalog No. 9004691EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
90-046-91EA 1 Ea.
1 options
Catalog No. 90-046-91EA Supplier Sigma Aldrich Supplier No. 9004691EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
These kits contain the following components:1. Maleimide gold nanoparticles (lyophilized).2. Ligand resuspension buffer.3. Reaction buffer.4. Quencher (lyophilized).Product of CytoDiagnostics, Inc.
Specifications
Quantity 1 Ea.
Synonym Au nanoparticles