Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 74194925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-194-925ML 25 mL
74-194-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-194-925ML Supplier Sigma Aldrich Supplier No. 74194925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles (AuNPs) are colloidal nanoparticles with multiple surface functionalities that can be used in the development of new biomaterials. AuNP has a spherical structure with a large surface to volume ratio and excellent biocompatibility. It is majorly used in a variety of biomedical applications ranging from diagnostics to drug delivery.Ideal for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications. Note: This product is not ideal for protein adsorption. For gold nanoparticles suitable for adsorption of proteins and other ligands for conjugate development, please see our gold nanoparticles stabilized in PBS.This material is highly monodisperse (<12% variability in size and shape), and provides significantly improved surface reactivity. Applications include Surface Enhanced Raman Lables, Sensing/Detection, Biological Targeting, Plasmonics and Electronics.Product of CytoDiagnostics, Inc.

Specifications

Quantity 25 mL
Molecular Formula Au
Linear Formula Au
MDL Number MFCD00003436
Synonym Au NP; Gold Colloid
Molecular Weight (g/mol) 196.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.