Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 75361025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-361-025ML 25 mL
75-361-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-361-025ML Supplier Sigma Aldrich Supplier No. 75361025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles (AuNPs), 20 nm are colloidal nanomaterials that have quasi-dynamic nature. Their properties include high stability, high surface area, good conductivity, and low toxicity. Their surface chemistry and biocompatibility allow them to be useful in biological applications.AuNPs can be used for a variety of applications such as biomedical, electrochemical and energy based devices. They can also be used in drug delivery and biosensing based device fabrication.Product of CytoDiagnostics, Inc.

Specifications

Quantity 25 mL
Molecular Formula Au
Linear Formula Au
MDL Number MFCD00003436
Synonym Au NP; Gold colloid
Molecular Weight (g/mol) 196.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.