Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanoparticles

Catalog No. 753637100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-363-7100ML 100 mL
75-363-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75-363-7100ML Supplier Sigma Aldrich Supplier No. 753637100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gold nanoparticles (AuNPs), 40 nm are quasi dynamic colloidal nanomaterials with good surface chemistry and high surface to volume ratio. Their properties include high stability, biocompatibility, and low toxicity which make them useful in biological applications.AuNPs can be used for a variety of applications such as biomedical, electrochemical and energy based devices. They can also be used in drug delivery and biosensing based device fabrication.Product of CytoDiagnostics, Inc.

Specifications

Molecular Formula Au
Linear Formula Au
Quantity 100 mL
MDL Number MFCD00003436
Synonym Au NP; Gold colloid
Molecular Weight (g/mol) 196.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.