Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gold nanorods

Catalog No. 905976175MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1.75 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-597-6175MG 1.75 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-597-6175MG Supplier Sigma Aldrich Supplier No. 9059761.75MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.Product of Nanopartz Inc.

Specifications

Quantity 1.75 mg
Molecular Formula Au
Linear Formula Au
MDL Number MFCD00003436
Synonym Cy3 labeled gold nanorods; Cy3 tagged gold nanorods; Dye-»labeled gold nanoparticles; Fluorescent-»labeled gold nanoparticles; Au nanorods; Gold nanorod
Molecular Weight (g/mol) 196.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.