Learn More
Sigma Aldrich Gold, nanorods
Catalog No. 90036525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
Catalog No. Quantity
90-036-525ML 25 mL
1 options
Catalog No. 90-036-525ML Supplier Sigma Aldrich Supplier No. 90036525ML
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Longitudinal peak = 1064 nm (+/- 15 nm)Longitudinal absorbance OD = 1Transverse peak = 508 nm (+/- 5 nm)Transverse absorbance OD = 0.1Citrate cappedOwing to the excellent optical properties arising from the surface plasmon resonance, gold nanorods find applications in biomedical imaging, drug delivery and photothermal treatment. The citrate capped nanorods are non-cytotoxic compared to the CTAB capped nanorods and are well suited for biomedical applications.This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922
Specifications
Quantity 25 mL
Synonym Au Nanorods