Learn More
Sigma Aldrich Gold nanospheres
Catalog No. 90563125MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
2.5 mg
Catalog No. Quantity
90-563-125MG 2.5 mg
1 options
Catalog No. 90-563-125MG Supplier Sigma Aldrich Supplier No. 9056312.5MG
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Owing to size and shape dependent quantum effects, the gold nanoparticles exhibit unique surface Plasmon resonance absorption, scattering, fluorescence, and photothermal properties which render them suitable for applications in catalysis, chemical sensing, biosensing , cellular and bioimaging, drug and gene delivery, and photothermal therapy. The fluorescent labeling of gold nanoparticles have been shown to cause emission enhancement in fluorophores. This makes the fluorescence tagged gold nanoparticles a dual mode nanoprobe for drug delivery and bioimaging applications.Product of Nanopartz Inc.
Specifications
Quantity 2.5 mg
Synonym Cy3 labeled gold nanospheres; Cy3 tagged gold nanospheres; Dye-»labeled gold nanoparticles; Fluorescent-»labeled gold nanoparticles