Learn More
Nanocomposix Ultra Uniform Gold Nanospheres - PEG-Carboxyl - 20 nm, 0.05 mg/mL in aq. 2 mM sodium citrate, 5 mL
SDP
Catalog No. NC1674764
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.20 nm Ultra Uniform spherical gold nanoparticles combine nearly perfect spherical shape, narrow size distribution, and strong plasmonic optical properties. These nanoparticles are useful in a broad range of applications from reference standards for size and particle number, biological labels, nanomedicine, drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC1674764 Supplier Nanocomposix Supplier No. AUXU205M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.20 nm Ultra Uniform spherical gold nanoparticles combine nearly perfect spherical shape, narrow size distribution, and strong plasmonic optical properties. These nanoparticles are useful in a broad range of applications from reference standards for size and particle number, biological labels, nanomedicine, drug delivery, biosensors, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, & UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.