Learn More
Electron Microscopy Sciences EMS Gold Palladium Targets(x3)
SDP
Catalog No. 5033104
Encompass
EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.   For use with the EMS 650 and 675X sputter coaters.
Catalog No. 50-331-04 Supplier Electron Microscopy Sciences Supplier No. 91031
EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.   For use with the EMS 650 and 675X sputter coaters.