Learn More
MilliporeSigma™ Supelco™ Gold-Plated Inlet Seals
Catalog No. 111008762 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Type:
Cross Design
Straight Design
Quantity:
10/Pk.
2/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
11-100-8762 Cross Design 10/Pk.
11-101-0666 Straight Design 2/Pk.
11-102-0768 Straight Design 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-8762 Supplier MilliporeSigma™ Supelco™ Supplier No. 23415U
Add to Cart
Add to Cart
Inlet seals on Agilent GCs must be regularly changed to prevent sample adsorption due to accumulation of sample residue and/or septum fragments
  • Replacement inlet seals are manufactured using high quality stainless steel plated with pure gold to insure inertness
  • No brighteners are used in the plating process
  • Each pack includes one washer for each seal
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Suitable for Applications with High Split Flows (> 200 mL/min.)
Includes One Washer for Each Seal
Material Stainless Steel
Quantity 10/Pk.
Description Gold-Plated
Type Cross Design