Description
- Replacement inlet seals are manufactured using high quality stainless steel plated with pure gold to insure inertness
- No brighteners are used in the plating process
- Each pack includes one washer for each seal
Specifications
Specifications
| For Use With (Application) | Suitable for Applications with Low Split Flows (< 200 mL/min.) |
| For Use With (Equipment) | Agilent GCs |
| Includes | One Washer for Each Seal |
| Material | Stainless Steel |
| Quantity | 10/Pk. |
| Description | Gold-Plated |
| Product Type | Straight Design |