Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold plating solution, electroless, metal content≈3.7g/L

Catalog No. AA42307K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42307K2 1 L
AA42307AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42307K2 Supplier Thermo Scientific Chemicals Supplier No. 042307K2

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

For coating nickel or copper

For coating nickel or copper

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold plating solution
Color Colorless to Yellow
Name Note Electroless, metal content ≈3.7g/L
MDL Number MFCD00003436
UN Number UN2922
Quantity 1 L
Physical Form Liquid
How should I prepare the surface of nickel or copper for gold plating with Gold plating solution, electroless, metal content≈3.7g/L?

Prior to plating, we recommend that you thoroughly clean the surface of the object to be plated. First, use a brush and a mild detergent to scrub the surface and then rinse thoroughly with purified water. Next, use a dilute hydrochloric (HCl) acid solution to remove any surface oxides and rinse thoroughly with purified water.

What instructions should I use for the Gold plating solution, electroless, metal content≈3.7g/L?

The gold plating solution can be kept in a glass beaker directly heated on a hot plate. The pH can drop with use, and we recommend adjusting the pH to the optimal range by adding small amounts of concentrated ammonium hydroxide.

What is the coverage of the Gold plating solution, electroless, metal content≈3.7g/L?

Assuming 50% of the gold is used, the coverage will be approximately 4096 cm2/L (635 in2/L or 2400 in2/gal) for an average thickness of 5 microinches of gold.

What is the deposition rate for the Gold plating solution, electroless, metal content≈3.7g/L?

The deposition rate for the Gold plating solution, electroless, metal content≈3.7g/L is 0.2-0.3 microinches/minute for a maximum thickness of 7-8 microinches in 30 minutes at 160°F (71°C). The thickness of gold can be increased to up to 15 microinches at a higher temperature of 175°F (80°C).

What are the operating conditions for the Gold plating solution, electroless, metal content≈3.7g/L?

The operating conditions for the Gold plating solution, electroless, metal content≈3.7 g/L are as follows:

  • pH 5.0-5.5
  • Agitation preferred
  • Time: at least 15 minutes
  • Temperature: 60-71°C (140-160°F)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.