Learn More
Perkin Elmer US LLC Gold Reflectance Mirror
Catalog No. 502113274
Encompass_Preferred
The PIKE 80Spec (L1272248) is a grazing angle specular reflection accessory that measures thin films and monolayers. Features include: A fixed 80° angle of incidence Polarizer mounts for optional IR polarizers Gold-coated reflective optics for highest energy throughput It is compatible with Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers
Catalog No. 50-211-3274 Supplier Perkin Elmer US LLC Supplier No. L1272377
The PIKE 80Spec (L1272248) is a grazing angle specular reflection accessory that measures thin films and monolayers. Features include: A fixed 80° angle of incidence Polarizer mounts for optional IR polarizers Gold-coated reflective optics for highest energy throughput It is compatible with Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers