Learn More
TA Instruments Gold Sample Pans, DSC Gold Pans, -180°C to 725°C, Use with Gold Lids, High Temperature Experiments, Standard DSC Cells, 10 Pack
Catalog No. 5000580733 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Gold Sample Pans are DSC gold pans useful for experiments over the temperature range of -180°C to 725°C. They are designed to be used with gold lids (P/N: 900868.901) for accurate thermal analysis in standard DSC cells.
Catalog No. 50-005-80733 Supplier TA Instruments Supplier No. 900866.901
Waters TA Instruments Gold Sample Pans are DSC gold pans useful for experiments over the temperature range of -180°C to 725°C. They are designed to be used with gold lids (P/N: 900868.901) for accurate thermal analysis in standard DSC cells.